Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEL D1 Major allergen 1 polypeptide chain 1 Rabbit anti-Feline, Polyclonal, FabGennix

Catalog No. 502206994
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6994 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6994 Supplier Fabgennix Inc Supplier No. FELD1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FEL D1 Major allergen 1 polypeptide chain 1 Polyclonal antibody specifically detects FEL D1 Major allergen 1 polypeptide chain 1 in Feline samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen FEL D1 Major allergen 1 polypeptide chain 1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene LOC677877
Gene Alias CH1; Major allergen I polypeptide chain 1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on feline Major Cat Allergen I polypeptide chain 1.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 677877
Target Species Feline
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.