Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline CCL5/RANTES Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF3819
This item is not returnable.
View return policy
Description
CCL5/RANTES Polyclonal specifically detects CCL5/RANTES in Feline samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
CCL5/RANTES | |
Polyclonal | |
Biotin | |
chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES | |
Sheep | |
Affinity Purified | |
RUO | |
6352 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CCL5 | |
E. coli-derived recombinant feline CCL5/RANTES | |
50 μg | |
Primary | |
Detects feline RANTES in ELISAs and Western blots. In sandwich immunoassays, less than 0.02% cross-reactivity with recombinant human RANTES, recombinant mouse RANTES, and recombinant cotton rat RANTES is observed. | |
Feline | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction