Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline CD4 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $514.50
Specifications
Antigen | CD4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Polyclonal |
Description
CD4 Polyclonal specifically detects CD4 in Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
CD4 | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Feline | |
920 | |
Mouse myeloma cell line NS0-derived recombinant feline CD4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects feline CD4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant canine CD4 is observed and 5% cross-reactivity with recombinant human CD4 and recombinant mouse CD4 is observed. |
LYOPH | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
CD4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title