Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline CD8 alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2598SP
This item is not returnable.
View return policy
Description
CD8 Polyclonal specifically detects CD8 in Feline samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CD8 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD8, CD8A, CD8a molecule, LEU2, p32 | |
Goat | |
Affinity Purified | |
RUO | |
925 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
P41688 | |
CD8A | |
Mouse myeloma cell line NS0-derived recombinant feline CD8 alpha Ala22-Tyr188 Accession # P41688 | |
25 μg | |
Primary | |
Detects feline CD8 alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse CD8 alpha is observed, approximately 5% cross-reactivity with recombinant canine CD8 alpha is observed, and less than 1% cross-reactivity with recombinant human CD8 alpha is observed. | |
Feline | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction