Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Fas/TNFRSF6/CD95 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2267
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2267 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2267 Supplier R&D Systems Supplier No. BAF2267
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Fas/TNFRSF6/CD95 Polyclonal specifically detects Fas/TNFRSF6/CD95 in Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL
Gene Accession No. NP_001009314
Gene Alias Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Gene Symbols FAS
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant feline Fas/TNFRSF6/CD95 Ala25-Lys172 Accession # NP_001009314
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity Detects feline Fas/TNFRSF6/CD95 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Fas and recombinant mouse Fas is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Feline
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.