Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline GM-CSF Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems BAM9873
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Feline samples. It is validated for ELISA Detection (Matched Antibody Pair).Specifications
GM-CSF | |
Monoclonal | |
LYOPH | |
ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
AAC06041 | |
CSF2 | |
E. coli-derived recombinant feline GM-CSF Ala18-Lys144 (Met36Ile, Thr56Ala & Lys126Asn) Accession # AAC06041 | |
250 μg | |
Primary | |
Detects feline GM-CSF in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant porcine, human, mouse, or rat GM-CSF was observed. | |
Feline | |
Purified |
ELISA | |
159319 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1437 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction