Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-1 beta/IL-1F2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1796
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Feline samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IL-1 beta/IL-1F2 | |
Polyclonal | |
Unconjugated | |
P41687 | |
IL1B | |
E. coli-derived recombinant feline IL-1 beta/IL-1F2 Ala116-Ser267 Accession # P41687 | |
100 μg | |
Primary | |
Detects IL-1 beta/IL-1F2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human IL-1 beta , recombinant mouse IL-1 beta , and recombinant canine IL-1 beta is observed. | |
Feline | |
IgG |
Western Blot, Neutralization, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.02-0.08 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Goat | |
Affinity Purified | |
RUO | |
3553 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction