Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB736SP
This item is not returnable.
View return policy
Description
IL-10 Monoclonal specifically detects IL-10 in Feline samples. It is validated for Western Blot, Neutralization.Specifications
IL-10 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3586 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Neutralization | |
137706 | |
Western Blot 1 ug/mL, Neutralization 0.2-0.6 ug/mL | |
P55029 | |
IL10 | |
E. coli-derived recombinant feline IL-10 Ser19-Ile178 Accession # P55029 | |
25 μg | |
Primary | |
Detects feline IL-10 in direct ELISAs and Western blots. In direct ELISAs, 40% cross-reactivity with canine IL-10 is observed and no cross-reactivity with IL-10 from human, mouse, rat, Epstein-Barr virus, or human cytomegalovirus is observed. | |
Feline | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction