Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-10 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | IL-10 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-10 Polyclonal specifically detects IL-10 in Feline samples. It is validated for Western Blot.Specifications
IL-10 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Feline | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
IL10 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P55029.2 | |
3586 | |
E. coli-derived recombinant feline IL-10 Ser19-Ile178 Accession # P55029.2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects feline IL-10 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant canine IL-10, recombinant human IL-10, and recombinant porcine IL-10 is observed and 10% cross-reactivity with recombinant mouse IL-10 and recombinant rat IL-10 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title