Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-18/IL-1F4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5989SP
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Polyclonal specifically detects IL-18/IL-1F4 in Feline samples. It is validated for Western Blot, Neutralization.Specifications
IL-18/IL-1F4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
Sheep | |
Affinity Purified | |
RUO | |
3606 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 2-12 ug/mL | |
Q95M33 | |
IL18 | |
E. coli-derived recombinant feline IL-18/IL-1F4 Tyr36-Asn192 Accession # Q95M33 | |
25 μg | |
Primary | |
Detects feline IL-18/IL-1F4 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant canine IL-18, approximately 30% cross-reactivity with recombinant porcine IL-18, approximately 20% cross-reactivity with recombinant human IL-18 and recombinant rat IL-18, and less than 10% cross-reactivity with recombinant mouse IL-18 and recombinant rhesus macaque IL-18 is observed. | |
Feline | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction