Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1890SP
This item is not returnable.
View return policy
Description
IL-2 Polyclonal specifically detects IL-2 in Feline samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IL-2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
Goat | |
Affinity Purified | |
RUO | |
3558 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.03-0.15 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q07885 | |
IL2 | |
E. coli-derived recombinant feline IL-2 Ala21-Thr154 (Cys146Ser) Accession # Q07885 | |
25 μg | |
Primary | |
Detects feline IL-2 in ELISAs and Western blots. In sandwich immunoassays, approximately 3% cross-reactivity with recombinant canine IL-2 is observed, approximately 1% cross reactivity with recombinant human IL-2 and recombinant porcine IL-2 is observed, and less than 0.2% cross-reactivity with recombinant mouse IL-2, recombinant equine IL-2, recombinant bovine IL-2, recombinant rat IL-2, and recombinant cotton rat IL-2 is observed. | |
Feline | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction