Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF984
This item is not returnable.
View return policy
Description
IL-4 Polyclonal specifically detects IL-4 in Feline samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-4 | |
Polyclonal | |
Biotin | |
P55030 | |
IL4 | |
E. coli-derived recombinant feline IL-4 (R&D Systems, Catalog # 984-FL) Gly24-His133 Accession # P55030 | |
50 μg | |
Primary | |
Detects feline IL-4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human IL-4, recombinant mouse IL-4, recombinant rat IL-4, recombinant canine IL-4, recombinant cotton rat IL-4, and recombinant porcine IL-4 is observed. | |
Feline | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Goat | |
Affinity Purified | |
RUO | |
3565 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction