Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline IL-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB23051
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB23051 100 μg
MAB23051SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB23051 Supplier R&D Systems Supplier No. MAB23051
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-6 Monoclonal specifically detects IL-6 in Feline samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen IL-6
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 341031
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41683
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Gene Symbols IL6
Host Species Mouse
Immunogen E. coli-derived recombinant feline IL-6 Thr28-Met208 (Glu133Lys) Accession # P41683
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3569
Test Specificity Detects recombinant feline IL-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-6, recombinant mouse (rm) IL-6, recombinant porcine IL-6, recombinant rat (rr) IL-6, recombinant canine IL-6, recombinant cotton rat IL-6, recombinant equine IL-6, rmIL-11, rmLIF, rmOSM, rhCLC or rrCNTF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Feline
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.