Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-8/CXCL8 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB22772
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Feline samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CXCL8/IL-8 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3576 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Neutralization, ELISA | |
344615 | |
Neutralization 0.075-0.375 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q9XSX5.1 | |
IL8 | |
E. coli-derived recombinant feline IL-8/CXCL8 Ala23-Ala101 Accession # Q9XSX5.1 | |
500 μg | |
Primary | |
Detects feline IL-8/CXCL8 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human IL-8/CXCL8 recombinant canine IL-8/CXCL8 or recombinant porcine IL-8/CXCL8 is observed. | |
Feline | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction