Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline IL-8/CXCL8 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2277SP
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Feline samples. It is validated for Western Blot, Neutralization.Specifications
CXCL8/IL-8 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 0.08-0.32 ug/mL | |
Q9XSX5.1 | |
IL8 | |
E. coli-derived recombinant feline IL-8/CXCL8 Ala23-Ala101 Accession # Q9XSX5.1 | |
25 μg | |
Primary | |
Detects feline IL-8/CXCL8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with over 75 tested chemokines of human, mouse, rat, porcine, or viral origin was observed. | |
Feline | |
Purified |
Western Blot, Neutralization | |
344609 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3576 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction