Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline Immunodeficiency Virus Antibody (5A1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064581AF750
Description
Feline Immunodeficiency Virus Monoclonal specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
Feline Immunodeficiency Virus | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Specific for feline immunodeficiency virus transmembrane protein gp41. It does not cross react with feline leukemia virus, feline herpes virus type 1, feline coronavirus or feline calicivirus. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
5A1 | |
Western Blot, ELISA | |
CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24 | |
Native, FIV extracts | |
Primary | |
Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction