Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Immunodeficiency Virus Antibody (5A1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199973 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017617 0.025 mg
NB10064581 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017617 Supplier Novus Biologicals Supplier No. NB100645810.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Immunodeficiency Virus Monoclonal specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Feline Immunodeficiency Virus
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5A1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Gene Alias CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24
Host Species Mouse
Immunogen Native, FIV extracts
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for feline immunodeficiency virus transmembrane protein gp41. It does not cross react with feline leukemia virus, feline herpes virus type 1, feline coronavirus or feline calicivirus.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.