Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline Immunodeficiency Virus Antibody (NM3-6A9), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066500MFV500
Description
Feline Immunodeficiency Virus Monoclonal antibody specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for Flow CytometrySpecifications
Feline Immunodeficiency Virus | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Flow Cytometry | |
NM3-6A9 | |
Flow Cytometry | |
CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24 | |
Native, extracts | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction