Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Feline Leukemia Virus Monoclonal Antibody (YCH)
GREENER_CHOICE

Catalog No. PIMA514707
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514707 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514707 Supplier Invitrogen™ Supplier No. MA514707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14707 targets Feline Leukemia Virus in ELISA, IP, and RIA applications and shows reactivity with Virus samples. The MA5-14707 immunogen is viral coat antigen. Product MA514707 is a smaller package size of MIF2801 (formerly sold as a Seradyn product).

Feline leukemia virus (FeLV) is a retrovirus that infects cats. As a retrovirus, FeLV is transmitted as an RNA virus but the RNA is reverse transcribed into DNA. Retroviruses contradict the Central dogma of molecular biology. FeLV is usually transmitted between infected cats when the transfer of saliva or nasal secretions is involved. If not defeated by the animal's immune system, the virus can be lethal. The disease caused by this virus is a form of cancer of blood cells called lymphocytes (a leukemia).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Feline Leukemia Virus
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone YCH
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene Alias FLV
Host Species Mouse
Immunogen Viral coat antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.