Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Feline Leukemia Virus P15E Monoclonal Antibody (PF6J-2A1)
GREENER_CHOICE

Catalog No. PIMA183883
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183883 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183883 Supplier Invitrogen™ Supplier No. MA183883
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blot, this antibody detects a band at ~15 and 20 kDa under reducing conditions. Mouse anti Feline leukemia virus p15e antibody, clone PF6J-2A1 recognizes an epitope within the amino-terminal domain of both the 20 kDa full-length (including intracellular sequence), and the 15k Da truncated forms, of feline leukaemia virus (FeLV) p15e transmembrane envelope protein.

Feline leukemia virus (FeLV) is a retrovirus that infects cats. As a retrovirus, FeLV is transmitted as an RNA virus but the RNA is reverse transcribed into DNA. Retroviruses contradict the Central dogma of molecular biology. FeLV is usually transmitted between infected cats when the transfer of saliva or nasal secretions is involved. If not defeated by the animal's immune system, the virus can be lethal. The disease caused by this virus is a form of cancer of blood cells called lymphocytes (a leukemia).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Feline Leukemia Virus P15E
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PF6J-2A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias FLV; FLV P15E
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.