Learn More
Invitrogen™ Feline Leukemia Virus p27 Monoclonal Antibody (7226)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547582
Description
Specificity: Specific for FeLv p27. Best pairs for p27 detection (capture/conjugate): Feline Leukemia Virus p27 Monoclonal antibody (Product # MA5-47582)/Feline Leukemia Virus p27 Monoclonal antibody (Product # MA1-7466). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Feline leukemia virus (FeLV) is a retrovirus that infects cats. As a retrovirus, FeLV is transmitted as an RNA virus but the RNA is reverse transcribed into DNA. Retroviruses contradict the Central dogma of molecular biology. FeLV is usually transmitted between infected cats when the transfer of saliva or nasal secretions is involved. If not defeated by the animal's immune system, the virus can be lethal. The disease caused by this virus is a form of cancer of blood cells called lymphocytes (a leukemia).
Specifications
Feline Leukemia Virus p27 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Lateral Flow, Immunocytochemistry | |
7226 | |
Unconjugated | |
FLV; FLV p27 | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.