Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Feline Leukemia Virus p27 Monoclonal Antibody (7228)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA547583

Catalog No. PIMA547583


Only null left
Add to Cart

Description

Description

Specificity: Specific for FeLv p27. Best pairs for p27 detection (capture/conjugate): Feline Leukemia Virus p27 Monoclonal antibody (Product # MA5-47583)/Feline Leukemia Virus p27 Monoclonal antibody (Product # MA1-7466); Feline Leukemia Virus p27 Monoclonal antibody (Product # MA5-47583)/Feline Leukemia Virus p27 Monoclonal antibody (Product # MA5-47583). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Feline leukemia virus (FeLV) is a retrovirus that infects cats. As a retrovirus, FeLV is transmitted as an RNA virus but the RNA is reverse transcribed into DNA. Retroviruses contradict the Central dogma of molecular biology. FeLV is usually transmitted between infected cats when the transfer of saliva or nasal secretions is involved. If not defeated by the animal's immune system, the virus can be lethal. The disease caused by this virus is a form of cancer of blood cells called lymphocytes (a leukemia).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Feline Leukemia Virus p27
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG1
ELISA, Lateral Flow, Immunocytochemistry
7228
Unconjugated
FLV; FLV p27
Proprietary sequence.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.