Learn More
Invitrogen™ Feline Leukemia Virus p27 Monoclonal Antibody (D67D)

Description
The MA1-7466 antibody reacts with Feline Leukemia virus FeLv p27 in viral and infected tissue samples. MA1-7466 has been successfully used in ELISA and Western blot applications. The MA1-7466 antibody was raised against FeLv p27 protein.
Specifications
Specifications
| Antigen | Feline Leukemia Virus p27 |
| Applications | ELISA, Lateral Flow, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D67D |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | FLV; FLV p27 |
| Host Species | Mouse |
| Immunogen | FeLv p27 protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.