Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Leukemia Virus P27 Monoclonal Antibody (PF12J-10A)
GREENER_CHOICE

Catalog No. MA183882
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183882 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183882 Supplier Thermo Scientific Supplier No. MA183882
Add to Cart
Add to Cart

Feline Leukemia Virus P27 Monoclonal Antibody (PF12J-10A)

Feline Leukemia Virus P27 Monoclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Feline Leukemia Virus P27
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PF12J-10A
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias FLV; FLV p27
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.