Learn More
Invitrogen™ Feline Leukemia Virus p27 Polyclonal Antibody, FITC

Description
The PA1-73180 antibody reacts with Feline Leukemia virus p27. PA1-73180 has been successfully used in Immunofluorescence applications. The PA1-73180 antibody was raised against purified FeLv p27.
Specifications
Specifications
| Antigen | Feline Leukemia Virus p27 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | FLV; FLV p27 |
| Host Species | Goat |
| Immunogen | Purified FeLv p27. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.