Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FEM1A Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518060
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
FEM1A (FEM-1 homolog a), also known as EPRAP (Prostaglandin E receptor 4-associated protein), is a 669 amino acid cytoplasmic protein that belongs to the FEM-1 family. FEM1A is present in atheromata and in macrophages derived from peripheral blood monocytes. FEM1A may be a component of an E3 ubiquitin-protein ligase complex and functions as a substrate recognition subunit. It is suggested that FEM1A participates in anti-inflammatory signaling of macrophages via its interaction with EP4. FEM1A is down-regulated in Rhabdomyosarcoma, the most common soft tissue neoplasm of children. FEM1A is considered a candidate gene for polycystic ovary syndrome.
Specifications
| FEM1A | |
| Polyclonal | |
| Unconjugated | |
| FEM1A | |
| AW611390; DKFZp762M136; EPRAP; fem-1 homolog A; fem-1 homolog a (C. elegans); Fem1a; Fem1aa; FEM1a-A; FEM1-alpha; FEM1-alpha-A; feminization 1 homolog a; feminization 1 homolog a (C. elegans); prostaglandin E receptor 4-associated protein; Protein fem-1 homolog A; protein fem-1 homolog A-A; similar to fem-1 homolog a (C.elegans) | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 55527 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BSK4 | |
| FEM1A | |
| Synthetic peptide sequence (GFIPEDLEAFIELH) corresponding to the C-terminus amino acids of FEM1A (aa 656-669). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction