Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEM1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550663 Supplier Invitrogen™ Supplier No. PA550663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total FEM1A protein.

FEM1A (FEM-1 homolog a), also known as EPRAP (Prostaglandin E receptor 4-associated protein), is a 669 amino acid cytoplasmic protein that belongs to the FEM-1 family. FEM1A is present in atheromata and in macrophages derived from peripheral blood monocytes. FEM1A may be a component of an E3 ubiquitin-protein ligase complex and functions as a substrate recognition subunit. It is suggested that FEM1A participates in anti-inflammatory signaling of macrophages via its interaction with EP4. FEM1A is down-regulated in Rhabdomyosarcoma, the most common soft tissue neoplasm of children. FEM1A is considered a candidate gene for polycystic ovary syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEM1A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FEM1A
Gene Accession No. Q9BSK4
Gene Alias AW611390; DKFZp762M136; EPRAP; fem-1 homolog A; fem-1 homolog a (C. elegans); Fem1a; Fem1aa; FEM1a-A; FEM1-alpha; FEM1-alpha-A; feminization 1 homolog a; feminization 1 homolog a (C. elegans); prostaglandin E receptor 4-associated protein; Protein fem-1 homolog A; protein fem-1 homolog A-A; similar to fem-1 homolog a (C.elegans)
Gene Symbols FEM1A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human fem-1 homolog a (C. elegans).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55527
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.