Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEM1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $485.00
Specifications
Antigen | FEM1B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FEM1B Polyclonal specifically detects FEM1B in Human samples. It is validated for Western Blot.Specifications
FEM1B | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DKFZp451E0710, F1AA, F1A-ALPHA, FEM-1 (C.elegans) homolog b, fem-1 homolog b (C. elegans), FEM1b, FEM1-beta, FEM-1-like death receptor binding protein, Fem-1-like death receptor-binding protein alpha, Fem-1-like in apoptotic pathway protein alpha, FIAA, KIAA0396, protein fem-1 homolog B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 368-627 of human FEM1B (NP_056137.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology | |
PBS with 50% glycerol, pH7.3. | |
10116 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title