Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEM1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111513 Supplier Invitrogen™ Supplier No. PA5111513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFMLQDRAKG LLGTTVTFDD LMGILCKSVL EIERAIKQTQ CPADPLQLNK ALSIILHLIC LLEKVPCTLE QDHFKKQTIY RFLKLHP Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 99%.

Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEM1C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FEM1C
Gene Accession No. Q96JP0
Gene Alias 2610312A07Rik; 3632443A22Rik; AI646567; EUROIMAGE686608; EUROIMAGE783647; fem-1 homolog C; fem-1 homolog c (C. elegans); fem-1 homolog c (C.elegans); FEM1A; FEM1c; FEM1-gamma; feminization 1 homolog a; feminization 1 homolog c; KIAA1785; Protein fem-1 homolog C; protein fem-1 homolog C pseudogene; protein fem-1 homolog C-like
Gene Symbols FEM1C
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FEM1C. Recombinant protein control fragment (Product #RP-107035).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56929
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.