Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEN-1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100320AF405
Description
FEN-1 Polyclonal antibody specifically detects FEN-1 in Human, Mouse samples. It is validated for Western BlotSpecifications
FEN-1 | |
Polyclonal | |
Western Blot | |
EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
The immunogen recognized by this antibody maps to a region between residues 75 and 125 of human Flap Structure Endonuclease 1 using the numbering given in entry NP_004102.1 (GeneID 2237). | |
0.1 mL | |
Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
2237 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction