Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FEN1 Recombinant Rabbit Monoclonal Antibody (3H9L9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535743
Description
Immunogen sequence: IQKHKSIEEI VRRLDPNKYP VPENWLHKEA HQLFLEPEVL DPESVELKWS EPNEEELIKF MCGEKQFSEE RIRSGVKRLS KSRQGSTQGR LDDFFKVTGS L.
FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
Specifications
FEN1 | |
Recombinant Monoclonal | |
0.23 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P39748, P39749 | |
FEN1 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human FEN-1 (P39748). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3H9L9 | |
Unconjugated | |
FEN1 | |
AW538437; DNase IV; fen1; FEN-1; fen1 {ECO:0000255; FEN-1 {ECO:0000255; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255; flap structure specific endonuclease 1; flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255; HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1; RAD2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14156, 2237 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction