Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588244 Supplier Invitrogen™ Supplier No. PA588244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDVLQNEEGE TTSHLMGMFY RTIRMMENGI KPVYVFDGKP PQLKSGELAK RSERRAEAEK QLQQAQAAGA EQEVEKFTKR LVKVTKQHND ECKHLLSLMG IPYLDAPSEA EASCAALVKA GKVYAAATED MDCLTFGSPV LMRHLTASEA KKLPIQEFHL SRILQELGLN QEQFVDLCIL LGSDYCESIR GIGPKRAVDL IQKHKSIEEI VRRLDPNKYP VPENWLHKEA HQLFLEPEVL DPESVELKWS EPNEEELIKF MCGEKQFSEE RIRSGVKRLS KSRQGSTQGR LDDFFKVTGS LSSAKRKEPE PKGSTKKKAK TGAAGKFKRG K.

FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEN1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FEN1
Gene Accession No. P39748, P39749
Gene Alias AW538437; DNase IV; fen1; FEN-1; fen1 {ECO:0000255; FEN-1 {ECO:0000255; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255; flap structure specific endonuclease 1; flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255; HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1; RAD2
Gene Symbols FEN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-380 of human FEN-1 (NP_0041021).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14156, 2237
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.