Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Fendiline Hydrochloride
GREENER_CHOICE

Catalog No. 640710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-071-0 10 mg
64-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-071-0 Supplier Tocris Bioscience™ Supplier No. 6407/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker

Fendiline Hydrochloride is a K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).

Specifications

Chemical Name or Material Ras GTPase Inhibitors
CAS 13636-18-5
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C23H25N.HCl
Formula Weight Observed MW: 351.91
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.