Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenhexamid 98.0+%, TCI America™
SDP

Catalog No. F09671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09671G 1 g
F0967200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09671G Supplier TCI America Supplier No. F09671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 126833-17-8
Molecular Formula C14H17Cl2NO2
Molecular Weight (g/mol) 302.20
MDL Number MFCD03095700
InChI Key VDLGAVXLJYLFDH-UHFFFAOYSA-N
Synonym fenhexamid, elevate, teldor, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexanecarboxamide, fenhexamide, decree, fenhexamid iso:bsi, unii-q68c3c9p1u, cyclohexanecarboxamide, n-2,3-dichloro-4-hydroxyphenyl-1-methyl, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexane-1-carboxamide
PubChem CID 213031
ChEBI CHEBI:81853
IUPAC Name N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide
SMILES CC1(CCCCC1)C(=O)NC1=CC=C(O)C(Cl)=C1Cl

Specifications

Melting Point 153°C
Boiling Point 320°C
Quantity 1 g
UN Number 3077
Formula Weight 302.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fenhexamid
RTECSNumber GU7879550
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.