Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fenitrothion Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP288845
Description
Fenitrothion Polyclonal specifically detects Fenitrothion in Non-species specific samples. It is validated for ELISA, Immunoaffinity Purification.Specifications
Fenitrothion | |
Polyclonal | |
ELISA 1:50000, Immunoaffinity Purification | |
agrothion, Cyfen | |
BSA-Fenitrothion Conjugate | |
0.1 mL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Affinity Purification | |
Unconjugated | |
Whole antiserum | |
Rabbit | |
Whole Antiserum | |
RUO | |
Non-species specific | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction