Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Fenobam
GREENER_CHOICESDP

Catalog No. 238650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-865-0 50 mg
23-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-865-0 Supplier R&D Systems™ Supplier No. 2386/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective mGlu5 negative allosteric modulator (NAM)

Potent and selective non-competitive mGlu5 negative allosteric modulator (NAM) that also displays inverse agonist properties; blocks mGlu5 constitutive activity in vitro (IC50 = 87 nM). Acts at an allosteric modulatory site shared with MPEP and binds the mGlu5 receptor with Kd values of 54 and 31 nM for rat and human receptors respectively. Displays anxiolytic activity following oral administration in vivo; also exhibits analgesic properties.

Chemical Identifiers

CAS 57653-26-6
Molecular Formula C11H11ClN4O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 266.69
Purity >99%
Molecular Weight (g/mol) 266.69
Chemical Name or Material Fenobam
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.