Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose octaacetate, 98%

Catalog No. AC419790050
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum

CAS: 126-14-7 | C28H38O19 | 678.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-14-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.59
InChI Key ZIJKGAXBCRWEOL-SAXBRCJISA-N
Synonym sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate
PubChem CID 31340
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point >78.0°C
Color Cream-White to White
Density 1.2800g/mL
Boiling Point 250.0°C (1.0 mmHg)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Merck Index 15,9013
Specific Gravity 1.28
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetic acid,acetone,benzene,90g/l in ethanol
Formula Weight 678.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sucrose octaacetate
Show More Show Less
EINECSNumber 204-772-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.