Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenoprefen calcium salt, Hydrate, MP Biomedicals™

Catalog No. ICN15803805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15803805 5 g
ICN15803810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15803805 Supplier MP Biomedicals, Inc Supplier No. 0215803805

Chemical Identifiers

CAS 53746-45-5
Molecular Formula C30H30CaO8
Molecular Weight (g/mol) 558.64
MDL Number MFCD00242722
InChI Key LZPBLUATTGKZBH-UHFFFAOYSA-L
Synonym fenopron, nalgesic, calcium-2-m-phenoxyphenyl propionate, hydrate, fenoprofen calcium salt, dl-2-3-phenoxyphenyl-propionic acid calcium salt, hydrate., fenoprofen calcium;phenyloxy brufen,calcium salt, 2-3-phenoxyphenyl propanoyl oxy calcio 2-3-phenoxyphenyl propanoate dihydrate, dihydrate 2-3-phenoxyphenyl propanoyl oxy calcio 2-3-phenoxyphenyl propanoate
PubChem CID 44119558
ChEBI CHEBI:5005
IUPAC Name calcium;2-(3-phenoxyphenyl)propanoate;dihydrate
SMILES CC(C1=CC(=CC=C1)OC2=CC=CC=C2)C(=O)[O-].CC(C1=CC(=CC=C1)OC2=CC=CC=C2)C(=O)[O-].O.O.[Ca+2]

Specifications

Color White
Quantity 5 g
Formula Weight 522.6
Physical Form Powder
Chemical Name or Material Fenoprofen Calcium Salt Hydrate
Recommended Storage Store at room temperature (15° to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.