Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FENS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145224 Supplier Invitrogen™ Supplier No. PA5145224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRVCDSCYDS IKDEDRTSLA TFHEGKHNIS HMSMDIARGL MVTCGTDRIV KI Antigen sequence identity: Mouse 98%; Rat 98%.

The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains a single FYVE domain and multiple WD40 repeats. This protein is localized to endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FENS1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDFY1
Gene Accession No. Q8IWB7
Gene Alias 1700013B03Rik; 1700120F24Rik; FENS1; FENS-1; FYVE domain-containing protein localized to endosomes 1; Jr1; KIAA1435; MGC137087 protein; mKIAA1435; Phosphoinositide-binding protein 1; phosphoinositide-binding protein SR1; WD repeat and FYVE domain containing 1; WD repeat and FYVE domain-containing protein 1; WD40 and FYVE domain containing 1; WD40- and FYVE domain-containing protein 1; WDF1; Wdfy1; ZFYVE17; zgc:101764; zgc:101764 protein; zgc:109909; zinc finger FYVE domain-containing protein 17
Gene Symbols WDFY1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 341-392. Recombinant protein control fragment (Product #RP-110069).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.