Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FENS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570319 Supplier Invitrogen™ Supplier No. PA570319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains a single FYVE domain and multiple WD40 repeats. This protein is localized to endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FENS1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WDFY1
Gene Accession No. E9Q4P1, Q8IWB7
Gene Alias 1700013B03Rik; 1700120F24Rik; FENS1; FENS-1; FYVE domain-containing protein localized to endosomes 1; Jr1; KIAA1435; MGC137087 protein; mKIAA1435; Phosphoinositide-binding protein 1; phosphoinositide-binding protein SR1; WD repeat and FYVE domain containing 1; WD repeat and FYVE domain-containing protein 1; WD40 and FYVE domain containing 1; WD40- and FYVE domain-containing protein 1; WDF1; Wdfy1; ZFYVE17; zgc:101764; zgc:101764 protein; zgc:109909; zinc finger FYVE domain-containing protein 17
Gene Symbols WDFY1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FENS1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57590, 69368
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.