Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fentanyl Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175148
Description
Working Concentration: 1/500. Sensitivity: 10 ng/mL Fentanyl produces 84% inhibition in a competitive ELISA employing Fentanyl polyclonal antibody.
Fentanyl is a synthetic, lipophilic phenylpiperidine opioid agonist with analgesic and anesthetic properties. Fentanyl selectively binds to the mu-receptor in the central nervous system (CNS) thereby mimicking the effects of endogenous opiates. Stimulation of the mu-subtype opioid receptor stimulates the exchange of GTP for GDP on the G-protein complex and subsequently inhibits adenylate cyclase. This results in a decrease in intracellular cAMP and leads to a reduction in the release of neurotransmitters such as substance P, GABA, dopamine, acetylcholine and noradrenaline. The analgesic effect of fentanyl is likely due to its metabolite morphine, which induces opening of G-protein-coupled inwardly rectifying potassium (GIRK) channels and blocks the opening of N-type voltage-gated calcium channels, thereby resulting in hyperpolarization and reduced neuronal excitability. [National Center for Biotechnology Information. PubChem Compound Database; CID=3345].
Specifications
Fentanyl | |
Polyclonal | |
Unconjugated | |
C22H28N2O; Duragesic; Fentanest; Fentanil; Phentanyl | |
Fentanyl-BSA. | |
1 mg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
13.26 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Sheep | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction