Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fer Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP120089AF350
Description
Fer Polyclonal specifically detects Fer in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
Fer | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated | |
EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Proto-oncogene c-Fer, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer | |
Rabbit | |
0.1 mL | |
2241 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
FER | |
Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-ProtNo. P16591] | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction