Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fer Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP120089APC
Description
Fer Polyclonal antibody specifically detects Fer in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, KnockDownSpecifications
Fer | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Knockdown Validated | |
EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Proto-oncogene c-Fer, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer | |
Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot# P16591] | |
0.1 mL | |
Protein Kinase | |
2241 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, KnockDown | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction