Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FER Monoclonal Antibody (1457CT181.12.17)
GREENER_CHOICE

Catalog No. PIMA537668
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537668 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537668 Supplier Invitrogen™ Supplier No. MA537668
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FER Monoclonal Antibody for IHC (P), Western Blot

FER is a member of the FPS/FES family of non-receptor tyrosine kinases. It regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors. FER is found in the cytoplasm and in the nucleus, where it was associated with the chromatin fraction. Its ubiquitous expression and its subcellular location indicate that it may be involved in key regulatory processes. The group IV cytoplasmic protein-tyrosine kinase Fer has been linked to cellular signaling responses to many different stimuli, including growth factors and cytokines. Data suggest a biological role for Fer in regulation of leukocyte recruitment during the innate immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FER
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1457CT181.12.17
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FER
Gene Accession No. P16591, P70451
Gene Alias AV082135; C330004K01Rik; c-FER; EC 2.7.10.2; Feline encephalitis virus-related kinase FER; FER; fer (fms/fps related) protein kinase; fer (fms/fps related) protein kinase, testis specific 2; fer (fps/fes related) tyrosine kinase; FER tyrosine kinase; Fert; Fert2; Flk; Flk_retired; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p47; p94-Fer; phosphoprotein NCP94; PPP1R74; protein phosphatase 1, regulatory subunit 74; proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase Fer; TYK3; Tyrosine kinase 3; tyrosine protein kinase FLK; tyrosine-protein kinase Fer; Tyrosine-protein kinase FLK
Gene Symbols FER
Host Species Mouse
Immunogen FER recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14158, 2241
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.