Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FERD3L Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen FERD3L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP180168
SDP
View Documents
Novus Biologicals
NBP180168
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

FERD3L Polyclonal specifically detects FERD3L in Human samples. It is validated for Western Blot.
Specifications

Specifications

FERD3L
Polyclonal
Rabbit
Human
Basic helix-loop-helix protein N-twist, BHLHA31, bHLHa31NTWIST, Class A basic helix-loop-helix protein 31, Fer3-like (Drosophila), fer3-like protein, MGC119861, NATO3Neuronal twist, Nephew of atonal 3, N-TWIST
FERD3L
IgG
19 kDa
Western Blot
Unconjugated
RUO
NP_690862
222894
Synthetic peptide directed towards the N terminal of human FERD3L. Peptide sequence SCVDTTVLDFVADLSLASPRRPLLCDFAPGVSLGDPALALREGRPRRMAR.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.