Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FERD3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541232 Supplier Invitrogen™ Supplier No. PA541232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SCVDTTVLDF VADLSLASPR RPLLCDFAPG VSLGDPALAL REGRPRRMAR Sequence homology: Human: 100%.

FERD3L is an evolutionarily conserved bHLH transcription factor that is identified in fly, mouse and man. Nervous system expression of this protein is detected in fly and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FERD3L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FERD3L
Gene Accession No. Q96RJ6
Gene Alias Basic helix-loop-helix protein N-twist; bHLHa31; Class A basic helix-loop-helix protein 31; fer3; Fer3-like (Drosophila); Fer3-like bHLH transcription factor; fer3-like protein; FERD3L; Mnato3; Nato; NATO3; nephew of atonal 3; Neuronal twist; NTWIST; N-TWIST
Gene Symbols FERD3L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FERD3L (aa 7-56).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222894
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.