Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FERD3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560389 Supplier Invitrogen™ Supplier No. PA560389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAYPESCVD TTVLDFVADL SLASPRRPLL CDFAPGVSLG DPALALREGR PRRMARFEEG DPEEEECEVD Q Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 56%.

FERD3L is an evolutionarily conserved bHLH transcription factor that is identified in fly, mouse and man. Nervous system expression of this protein is detected in fly and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FERD3L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FERD3L
Gene Accession No. Q96RJ6
Gene Alias Basic helix-loop-helix protein N-twist; bHLHa31; Class A basic helix-loop-helix protein 31; fer3; Fer3-like (Drosophila); Fer3-like bHLH transcription factor; fer3-like protein; FERD3L; Mnato3; Nato; NATO3; nephew of atonal 3; Neuronal twist; NTWIST; N-TWIST
Gene Symbols FERD3L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FERD3L. Recombinant protein control fragment (Product #RP-98483).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.