Learn More
Description
5-Fluoroorotic Acid is a yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3), and E. coli (pyrF).
Recommended for:
Detection of URA3 gene expression in yeast; Construction of yeast 2-hybrid libraries (1); Other areas of yeast genetics including gene replacement, analysis of transposable genetic elements and identification of gene mutations.
Note:
Low solubility in water. Solubility in dimethylsulfoxide is greater than 150mg/mL. Preparation of a 100mg/mL solution in dime-thylsulfoxide is recommended.
Specifications
Specifications
| Chemical Name or Material | 5-Fluoroorotic Acid |
| Product Type | 5-Fluoroorotic Acid |
| Quantity | 1 g |
| Form | Powder |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.