Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 5-Fluoroorotic Acid

Catalog No. p-4530619
Click to view available options
Quantity:
1 g
5 g

Catalog No. FERR0811

Supplier: Thermo Scientific™ R0811

Only null left
Add to Cart
Add to Cart

Detect expression of the URA3 gene, which encodes orotine-5'-monophosphate (OMP) dicarboxylase with Thermo Scientific™ 5-Fluoroorotic Acid, used in yeast molecular genetics.

5-Fluoroorotic Acid is a yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3), and E. coli (pyrF).

Yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3) and E.coli (pyrF).

Recommended for:

Detection of URA3 gene expression in yeast; Construction of yeast 2-hybrid libraries (1); Other areas of yeast genetics including gene replacement, analysis of transposable genetic elements and identification of gene mutations.

Note:

Low solubility in water. Solubility in dimethylsulfoxide is greater than 150mg/mL. Preparation of a 100mg/mL solution in dime-thylsulfoxide is recommended.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluoroorotic Acid
Product Type 5-Fluoroorotic Acid
Quantity 1 g
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.