Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DEPC-treated Water
GSA_VA

Catalog No. p-4530715
Click to view available options
Quantity:
30 mL
5 x 1 mL

Optimize applications involving RNA with this deionized, diethylpyrocarbonate (DEPC) treated and 0.22μm membrane-filtered water.

deionized, diethylpyrocarbonate (DEPC) is treated and 0.22 μm membrane-filtered water. It is ideal for applications involving RNA.

Ideal for applications involving RNA.
  • Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases

Note:

DEPC-treated Water is not recommended for PCR or in vivo experiments (1).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

pH 7.9
Grade Molecular Biology
Packaging Vial
Treatment(s) DEPC-Treated
Quantity 5 x 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.